Learn More
ABclonal Technology CYP3A4 Rabbit mAb
SDP
Catalog No. 5000005941
Encompass_Preferred
This gene encodes a member of the cytochrome P450 superfamily of enzymes The cytochrome P450 proteins are monooxygenases that catalyze many reactions involved in drug metabolism and synthesis of cholesterol steroids and other lipids This protein localizes to the endoplasmic reticulum and its expression is induced by glucocorticoids and some pharmacological agents This enzyme is involved in the metabolism of approximately half the drugs in use today including acetaminophen codeine cyclosporin A diazepam erythromycin and chloroquine The enzyme also metabolizes some steroids and carcinogens This gene is part of a cluster of cytochrome P450 genes on chromosome 7q21 1 Previously another CYP3A gene CYP3A3 was thought to exist however it is now thought that this sequence represents a transcript variant of CYP3A4 Alternatively spliced transcript variants encoding different isoforms have been identified
Catalog No. 50-000-05941 Supplier ABclonal Technology Supplier No. A22229100UL
This gene encodes a member of the cytochrome P450 superfamily of enzymes The cytochrome P450 proteins are monooxygenases that catalyze many reactions involved in drug metabolism and synthesis of cholesterol steroids and other lipids This protein localizes to the endoplasmic reticulum and its expression is induced by glucocorticoids and some pharmacological agents This enzyme is involved in the metabolism of approximately half the drugs in use today including acetaminophen codeine cyclosporin A diazepam erythromycin and chloroquine The enzyme also metabolizes some steroids and carcinogens This gene is part of a cluster of cytochrome P450 genes on chromosome 7q21 1 Previously another CYP3A gene CYP3A3 was thought to exist however it is now thought that this sequence represents a transcript variant of CYP3A4 Alternatively spliced transcript variants encoding different isoforms have been identified