Learn More
Aviva Systems Biology Corp CYP3A43 Antibody - middle region (ARP49757_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP49757_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CYP3A43. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human CYP3A43. Alternate names: MGC119315, MGC119316ApplicationWB, IHC. Species reactivity: Cow, Dog, Guinea Pig, Human, Rabbit, Rat, Sheep
Catalog No. 50-014-60171