Learn More
Aviva Systems Biology Corp CYP3A43 Antibody - C-terminal region (ARP49758_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP49758_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CYP3A43. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human CYP3A43. Alternate names: MGC119315, MGC119316ApplicationWB, IHC. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Sheep
Catalog No. 50-014-60172