Learn More
Aviva Systems Biology Corp CYP3A5 Antibody - N-terminal region (ARP60135_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP60135_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CYP3A5. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N terminal region of human CYP3A5. Alternate names: CP35, P450PCN3, PCN3, CYPIIIA5ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rat, Sheep
Catalog No. 50-286-955