Learn More
Aviva Systems Biology Corp CYP3A7 Antibody - middle region (ARP41801_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP41801_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against CYP3A7. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human CYP3A7. Alternate names: CP37, P450-HFLA, CYPIIIA7ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Human, Rat
Catalog No. 50-014-57997