Learn More
Medchemexpress LLC CYP3cide (PF-4981517) | 1390637-82-7 | 99.9% | 456.59 | 5 MG
SDP
Catalog No. 5000388194
Encompass_Preferred
CYP3cide (PF-4981517) is a potent, selective, and time-dependent inhibitor of cytochrome P4503A4 (CYP3A4). It can be used to distinguish the contributions of CYP3A4 versus CYP3A5 on agent metabolism. The IC50 values for Midazolam 1'-hydroxylase activity are 0.03 μM, 17 μM, and 71 μM for CYP3A4, CYP3A5, and CYP3A7, respectively.
  • Potent, selective, and time-dependent inhibitor of CYP3A4.
  • Useful for distinguishing the roles of CYP3A4 and CYP3A5 in drug metabolism.
  • Purity: 99.92%.
  • Available in solid and solution forms.
Catalog No. 50-003-88194 Supplier Medchemexpress LLC Supplier No. HY186425MG
CYP3cide (PF-4981517) is a potent, selective, and time-dependent inhibitor of cytochrome P4503A4 (CYP3A4). It can be used to distinguish the contributions of CYP3A4 versus CYP3A5 on agent metabolism. The IC50 values for Midazolam 1'-hydroxylase activity are 0.03 μM, 17 μM, and 71 μM for CYP3A4, CYP3A5, and CYP3A7, respectively.
  • Potent, selective, and time-dependent inhibitor of CYP3A4.
  • Useful for distinguishing the roles of CYP3A4 and CYP3A5 in drug metabolism.
  • Purity: 99.92%.
  • Available in solid and solution forms.