Learn More
Aviva Systems Biology Corp CYP4A22 Antibody - N-terminal region (ARP41862_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP41862_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CYP4A22. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human CYP4A22. Alternate names: -ApplicationWB. Species reactivity: Human
Catalog No. 50-014-58021