Learn More
Aviva Systems Biology Corp CYP4F11 Antibody - N-terminal region (ARP42588_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP42588_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against CYP4F11. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human CYP4F11. Alternate names: CYPIVF11ApplicationWB. Species reactivity: Cow, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-58275