Learn More
Aviva Systems Biology Corp CYP4F2 Antibody - C-terminal region (ARP63725_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP63725_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CYP4F2. It was validated on Western Blot. . Alternate names: CPF2ApplicationWB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Sheep
Catalog No. 50-288-064