Learn More
Aviva Systems Biology Corp CYP7B1 Antibody - C-terminal region (ARP60826_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP60826_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CYP7B1. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C terminal region of human CYP7B1. Alternate names: CBAS3, CP7B, SPG5AApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Pig, Rabbit, Rat
Catalog No. 50-287-183