Learn More
Aviva Systems Biology Corp CYP8B1 Antibody - middle region (ARP60138_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP60138_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CYP8B1. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of human CYP8B1. Alternate names: CP8B, CYP12, FLJ17826ApplicationWB. Species reactivity: Human
Catalog No. 50-286-956