Learn More
Enzo Life Sciences Cypermethrin (50mg). CAS: 52315-07-8
SDP
Supplier:  Enzo Life Sciences BMLPR1000050
Encompass_Preferred
Synthetic type II pyrethroid. Potent calcineurin (protein phosphatase 2B) inhibitor IC50~0.03 nM. May also have varied effects on ion channels. Serious doubts have been raised about the efficacy of this inhibitor. Alternative name: Alphamethrine, Pestanal, (R,S)-alpha-Cyano-3-phenoxybenzyl(1RS)-cis,trans-3-(2,2-dichlorovinyl)-2,2-dimethylcyclopropane carboxylate. Purity: ≥96% (HPLC) (mixture of isomers:racemic and cis/trans). Solubility: Soluble in ethanol (25mg/ml). Insoluble in water. Long Term Storage: Ambient.
Catalog No. 50-201-1498