Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cypermethrin, 95%, Thermo Scientific™

Catalog No. AAJ6599803
Catalog No. AAJ6599803 Supplier Thermo Scientific Chemicals Supplier No. J6599803

missing translation for 'validMainframeMsg'

Potent inhibitor of calcineurin; a pyrethrin

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Cypermethrin is used as pharmacologically active substances and their classification regarding maximum residue limits in foodstuffs of animal origin.

Solubility
Cypermethrin is soluble in methanol or acetone. The product is insoluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Specifications

CAS 52315-07-8
Flash Point 100°C (212°F)
Molecular Formula C22H19Cl2NO3
MDL Number MFCD00055328
UN Number UN2811
Merck Index 14,2767
Synonym alpha-Cyano-(3-phenoxyphenyl)methyl 3-(2,2-dichlorovinyl)-2,2-dimethylcyclopropanecarboxylate
InChI Key KAATUXNTWXVJKI-UHFFFAOYSA-N
SMILES CC1(C(C1C(=O)OC(C#N)C2=CC(=CC=C2)OC3=CC=CC=C3)C=C(Cl)Cl)C
IUPAC Name [cyano-(3-phenoxyphenyl)methyl] 3-(2,2-dichloroethenyl)-2,2-dimethylcyclopropane-1-carboxylate
Molecular Weight (g/mol) 416.298
PubChem CID 2912
ChEBI CHEBI:4042
Formula Weight 416.3
Percent Purity 95%
Chemical Name or Material Cypermethrin
Show More Show Less
Hazard Category Warning
Hazard Statement GHS H Statement
H302-H332-H335
Harmful if swallowed.
Harmful if inhaled.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P261-P304+P340-P301+P312-P312-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Call a POISON CENTER or doctor/physician if you feel unwell.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
RTECSNumber GZ1250000
Recommended Storage −30° to −10°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.