Learn More
Cayman Chemical CypermethrIn-d5 2.5mg
SDP
Catalog No. 502916886
Encompass_Preferred
An internal standard for the quantification of cypermethrin by GC- or LC-MS
Catalog No. 50-291-6886 Supplier Cayman Chemical Supplier No. 392652.5 MG
An internal standard for the quantification of cypermethrin by GC- or LC-MS