Learn More
Cayman Chemical Cypermethrin-d9, Purity: ≥99% deuterated forms (d1-d9), 5 mg
SDP
Catalog No. 502915733
Encompass_Preferred
An internal standard for the quantification of cypermethrin by GC- or LC-MS
Catalog No. 50-291-5733 Supplier Cayman Chemical Supplier No. 392665 MG
An internal standard for the quantification of cypermethrin by GC- or LC-MS