Learn More
Abcam CyPPA, SK channel activator, 10MG
Catalog No. 502707218
Encompass_Preferred
MW 285.39 g/mol, Purity >98%. First selective small conductance Ca²+-activated K+ channel (SK) activator. Selectively opens SK2 and SK3 channels (EC₅₀ values are 5.6 and 14 μM, respectively). Shows no activity at SK1 and IK. Additionally reduces activity and excitability of dopamine neurons. Active in vivo.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-7218 Supplier Abcam Supplier No. AB12082910MG
MW 285.39 g/mol, Purity >98%. First selective small conductance Ca²+-activated K+ channel (SK) activator. Selectively opens SK2 and SK3 channels (EC₅₀ values are 5.6 and 14 μM, respectively). Shows no activity at SK1 and IK. Additionally reduces activity and excitability of dopamine neurons. Active in vivo.
The product is subject to the following: Abcam Restricted Use Statement