Learn More
Cayman Chemical CyprIdIna LucIfRIntrIfluor
SDP
Catalog No. 502942914
Encompass_Preferred
A chemiluminescent substrate of luciferase; upon hydrolysis by luciferase, em = 465 nm; has been used in ERα reporter assays for the discovery of ERα-activating compounds in vitro and in BRET assays for tumor detection in vivo
Catalog No. 50-294-2914 Supplier Cayman Chemical Supplier No. 41088500 UG
A chemiluminescent substrate of luciferase; upon hydrolysis by luciferase, em = 465 nm; has been used in ERα reporter assays for the discovery of ERα-activating compounds in vitro and in BRET assays for tumor detection in vivo