Learn More
Cayman Chemical Cypridina Luciferin (trifluoroacetate salt), Purity: ≥95%, 1 mg
SDP
Catalog No. 502942913
Encompass_Preferred
A chemiluminescent substrate of luciferase; upon hydrolysis by luciferase, em = 465 nm; has been used in ERα reporter assays for the discovery of ERα-activating compounds in vitro and in BRET assays for tumor detection in vivo
Catalog No. 50-294-2913 Supplier Cayman Chemical Supplier No. 410881 MG
A chemiluminescent substrate of luciferase; upon hydrolysis by luciferase, em = 465 nm; has been used in ERα reporter assays for the discovery of ERα-activating compounds in vitro and in BRET assays for tumor detection in vivo