Learn More
Cayman Chemical Cyproconazole, Purity: ≥98% (mixture of isomers), CAS: 94361-06-5, 5 mg
SDP
Catalog No. 502950982
Encompass_Preferred
A triazole fungicide; reduces mycelial growth of A. mellea on grapevine roots; reduces defoliation of rose plants induced by D. rosae; Lethal to C. riparius and D. tigrina (LC50s = 17.46 and 47.38 mg/L in tank water, respectively); induces liver hypertrophy and fat vacuolation, as well as induces hepatocellular adenoma and carcinoma formation, in mice
Catalog No. 50-295-0982 Supplier Cayman Chemical Supplier No. 329475 MG
A triazole fungicide; reduces mycelial growth of A. mellea on grapevine roots; reduces defoliation of rose plants induced by D. rosae; Lethal to C. riparius and D. tigrina (LC50s = 17.46 and 47.38 mg/L in tank water, respectively); induces liver hypertrophy and fat vacuolation, as well as induces hepatocellular adenoma and carcinoma formation, in mice