Learn More
Medchemexpress LLC Cyproconazole (Standard) | 94361-06-5 | 98.2% | C15H18ClN3O | 250 MG
SDP
Catalog No. 5000357613
Encompass_Preferred
Cyproconazole (Standard) is an analytical standard intended for research and analytical applications. This triazole fungicide protects crops, fruits, and vegetables against a wide range of fungal pathogens. It has been observed to cause hepatocellular adenomas and carcinomas in CD-1 mice, and exhibits low toxicity to zebrafish embryos, affecting locomotor activity.
  • Analytical standard for reference assays
  • Used in qualitative, quantitative, and methodological research experiments
  • Suitable for HPLC, GC, and MS applications
Catalog No. 50-003-57613 Supplier Medchemexpress LLC Supplier No. 250MGHYA0277R
Cyproconazole (Standard) is an analytical standard intended for research and analytical applications. This triazole fungicide protects crops, fruits, and vegetables against a wide range of fungal pathogens. It has been observed to cause hepatocellular adenomas and carcinomas in CD-1 mice, and exhibits low toxicity to zebrafish embryos, affecting locomotor activity.
  • Analytical standard for reference assays
  • Used in qualitative, quantitative, and methodological research experiments
  • Suitable for HPLC, GC, and MS applications