Learn More
Medchemexpress LLC Cyproconazole (Standard) | 94361-06-5 | 98.7% | C15H18ClN3O | 500 MG
SDP
Catalog No. 5000432577
Encompass_Preferred
Cyproconazole is a triazole fungicide effective against a wide range of fungal pathogens in crops, fruits, and vegetables. Research indicates it can induce hepatocellular adenomas and carcinomas in CD-1 mice and exhibits low toxicity to zebrafish embryos, impacting locomotor activity.
  • Fungicide that protects crops, fruits, and vegetables
  • Effective against a wide range of fungal pathogens
  • Induces time- and dose-dependent mortality in zebrafish embryos
  • Inhibits spontaneous movement in zebrafish embryos
  • Increases pericardial edema and cumulative deformity rate in zebrafish embryos/larvae
  • Decreases basal respiration and oligomycin-induced ATP respiration in zebrafish embryos
  • Increases hepatocellular proliferation in male mice
  • Causes a larger and more sustained decrease in plasma cholesterol levels
  • Affects fat deposition in the liver
Catalog No. 50-004-32577 Supplier Medchemexpress LLC Supplier No. HYA0277500MG
Cyproconazole is a triazole fungicide effective against a wide range of fungal pathogens in crops, fruits, and vegetables. Research indicates it can induce hepatocellular adenomas and carcinomas in CD-1 mice and exhibits low toxicity to zebrafish embryos, impacting locomotor activity.
  • Fungicide that protects crops, fruits, and vegetables
  • Effective against a wide range of fungal pathogens
  • Induces time- and dose-dependent mortality in zebrafish embryos
  • Inhibits spontaneous movement in zebrafish embryos
  • Increases pericardial edema and cumulative deformity rate in zebrafish embryos/larvae
  • Decreases basal respiration and oligomycin-induced ATP respiration in zebrafish embryos
  • Increases hepatocellular proliferation in male mice
  • Causes a larger and more sustained decrease in plasma cholesterol levels
  • Affects fat deposition in the liver