Learn More
Medchemexpress LLC Cyprodinil 100mg | 121552-61-2 | 100 MG
SDP
Catalog No. 5000227963
Encompass_Preferred
Cyprodinil is an anilinopyrimidine fungicide supplied as an analytical standard for laboratory research and analytical applications. It is provided as a solid reference material (100 mg) with high purity for use in method development, calibration, and quality control; not for human use.
  • Analytical standard for research and method development.
  • High purity (≈98.8%), suitable for reference and quality control.
  • Solid form supplied as a 100 mg vial for convenient handling.
  • Used as a fungicide reference in residue and environmental analysis.
  • Store under recommended conditions to maintain stability.
Catalog No. 50-002-27963 Supplier Medchemexpress LLC Supplier No. HY116214100MG
Cyprodinil is an anilinopyrimidine fungicide supplied as an analytical standard for laboratory research and analytical applications. It is provided as a solid reference material (100 mg) with high purity for use in method development, calibration, and quality control; not for human use.
  • Analytical standard for research and method development.
  • High purity (≈98.8%), suitable for reference and quality control.
  • Solid form supplied as a 100 mg vial for convenient handling.
  • Used as a fungicide reference in residue and environmental analysis.
  • Store under recommended conditions to maintain stability.