Learn More
Medchemexpress LLC Cyprodinil | 121552-61-2 | 98.8% | 225.29 | 50 MG
SDP
Catalog No. 5000364118
Encompass_Preferred
Cyprodinil (CGA-219417) is a broad-spectrum anilinopyrimidine fungicide known for its anti-androgenic and androgenic activities. It functions by inhibiting methionine biosynthesis in plant-pathogenic fungi, offering protection to fruits and vegetables from various pathogens. This compound also acts as an activator of the aryl hydrocarbon receptor.
  • Inhibits methionine biosynthesis in plant-pathogenic fungi
  • Protects fruits and vegetables from various pathogens
  • Exhibits anti-androgenic and androgenic activities
  • Activator of the aryl hydrocarbon receptor
  • Increases protein expression of CYP1A1
  • Induces phosphorylation of ERK1/2
Catalog No. 50-003-64118 Supplier Medchemexpress LLC Supplier No. 50MGHY116214
Cyprodinil (CGA-219417) is a broad-spectrum anilinopyrimidine fungicide known for its anti-androgenic and androgenic activities. It functions by inhibiting methionine biosynthesis in plant-pathogenic fungi, offering protection to fruits and vegetables from various pathogens. This compound also acts as an activator of the aryl hydrocarbon receptor.
  • Inhibits methionine biosynthesis in plant-pathogenic fungi
  • Protects fruits and vegetables from various pathogens
  • Exhibits anti-androgenic and androgenic activities
  • Activator of the aryl hydrocarbon receptor
  • Increases protein expression of CYP1A1
  • Induces phosphorylation of ERK1/2