Learn More
Medchemexpress LLC Cyprodinil-d5 5mg | 1773496-67-5 | 230.32 | C14H10D5N3 | 5 MG
SDP
Catalog No. 5000227307
Encompass_Preferred
Cyprodinil-d5 is the deuterium-labeled isotopologue of cyprodinil supplied as an analytical reference standard for laboratory use. It is intended for use as an internal standard or tracer in mass spectrometry, residue analysis, and metabolic studies where isotopic labeling improves quantification and compound identification. The material is provided in small milligram-scale packages appropriate for analytical workflows.
  • Deuterium-labeled internal standard for analytical quantification.
  • Useful for mass spectrometry and residue analysis.
  • Chemical formula: C14H10D5N3; molecular weight: 230.32.
  • Offered in small milligram packages for laboratory workflows.
  • Intended for research use only.
Catalog No. 50-002-27307 Supplier Medchemexpress LLC Supplier No. HY116214S5MG
Cyprodinil-d5 is the deuterium-labeled isotopologue of cyprodinil supplied as an analytical reference standard for laboratory use. It is intended for use as an internal standard or tracer in mass spectrometry, residue analysis, and metabolic studies where isotopic labeling improves quantification and compound identification. The material is provided in small milligram-scale packages appropriate for analytical workflows.
  • Deuterium-labeled internal standard for analytical quantification.
  • Useful for mass spectrometry and residue analysis.
  • Chemical formula: C14H10D5N3; molecular weight: 230.32.
  • Offered in small milligram packages for laboratory workflows.
  • Intended for research use only.