Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Cyprodinil Standard 10mg | 121552-61-2 | 225.29 g/mol | C14H15N3 | 10 MG
SDP

Catalog No. 5000229175
Encompass_Preferred

Cyprodinil analytical standard for use in analytical testing, method development, calibration, and quality control. Supplied as a precisely weighed solid suitable for preparing calibration and reference solutions.

  • Analytical standard grade for method development and quality control.
  • High purity: 99.04% (supplied assay).
  • CAS number: 121552-61-2 for unambiguous identification.
  • Available in small pack sizes for trace-level calibration (example: 10 MG).
  • Molecular formula C14H15N3 and molecular weight 225.29 g/mol.

Catalog No. 50-002-29175 Supplier Medchemexpress LLC Supplier No. HY116214R10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Cyprodinil analytical standard for use in analytical testing, method development, calibration, and quality control. Supplied as a precisely weighed solid suitable for preparing calibration and reference solutions.

  • Analytical standard grade for method development and quality control.
  • High purity: 99.04% (supplied assay).
  • CAS number: 121552-61-2 for unambiguous identification.
  • Available in small pack sizes for trace-level calibration (example: 10 MG).
  • Molecular formula C14H15N3 and molecular weight 225.29 g/mol.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.