Learn More
Medchemexpress LLC Cyprodinil Standard 25mg | 121552-61-2 | 225.29 g/mol | C14H15N3 | 25 MG
SDP
Catalog No. 5000227128
Encompass_Preferred
Cyprodinil (Standard) is an analytical reference standard of the fungicide cyprodinil (CAS 121552-61-2) supplied as a small, stable solid for research and analytical use. The product is provided as a 25 mg unit of high-purity material (99.04%) with confirmed molecular formula C14H15N3 and molecular weight 225.29 g/mol, intended for calibration, method development, and quality-control workflows in chromatography and mass spectrometry.
  • High purity (99.04%) suitable for analytical applications.
  • Confirmed molecular formula C14H15N3 and molecular weight 225.29 g/mol.
  • Intended for calibration, method development, and method validation.
  • Appropriate for pesticide residue testing and environmental analysis.
  • Supplied as a stable solid convenient for storage and handling.
Catalog No. 50-002-27128 Supplier Medchemexpress LLC Supplier No. HY116214R25MG
Cyprodinil (Standard) is an analytical reference standard of the fungicide cyprodinil (CAS 121552-61-2) supplied as a small, stable solid for research and analytical use. The product is provided as a 25 mg unit of high-purity material (99.04%) with confirmed molecular formula C14H15N3 and molecular weight 225.29 g/mol, intended for calibration, method development, and quality-control workflows in chromatography and mass spectrometry.
  • High purity (99.04%) suitable for analytical applications.
  • Confirmed molecular formula C14H15N3 and molecular weight 225.29 g/mol.
  • Intended for calibration, method development, and method validation.
  • Appropriate for pesticide residue testing and environmental analysis.
  • Supplied as a stable solid convenient for storage and handling.