Learn More
Cayman Chemical Cyproheptadine-d3, Purity: ≥99% deuterated forms (d1-d3), CAS: 2712455-05-3, 1 mg
SDP
Catalog No. 502941371
Encompass_Preferred
An internal standard for the quantification of cyproheptadine by GC- or LC-MS
Catalog No. 50-294-1371 Supplier Cayman Chemical Supplier No. 290231 MG
An internal standard for the quantification of cyproheptadine by GC- or LC-MS