Learn More
Medchemexpress LLC Cyproheptadine hydrochloride | 969-33-5 | 99.3% | 1 ML
SDP
Catalog No. 5000407312
Encompass_Preferred
Cyproheptadine hydrochloride is a potent and orally active 5-HT2A receptor antagonist. It exhibits antidepressant, antiserotonergic, antiplatelet, and thromboprotective activities. This product is suitable for research into thromboembolic disorders and other related conditions.
  • Potent 5-HT2A receptor antagonist
  • Orally active compound
  • Exhibits antidepressant and antiserotonergic effects
  • Shows antiplatelet and thromboprotective activities
  • For research use only
Catalog No. 50-004-07312 Supplier Medchemexpress LLC Supplier No. HYB0366A10MM/1ML
Cyproheptadine hydrochloride is a potent and orally active 5-HT2A receptor antagonist. It exhibits antidepressant, antiserotonergic, antiplatelet, and thromboprotective activities. This product is suitable for research into thromboembolic disorders and other related conditions.
  • Potent 5-HT2A receptor antagonist
  • Orally active compound
  • Exhibits antidepressant and antiserotonergic effects
  • Shows antiplatelet and thromboprotective activities
  • For research use only