Learn More
Medchemexpress LLC Cyproheptadine hydrochloride sesquihydrate | 41354-29-4 | 99.7% | 1 ML
SDP
Catalog No. 5000412354
Encompass_Preferred
Cyproheptadine hydrochloride sesquihydrate is an antihistamine and an antagonist of serotonin and histamine2.
  • Antihistamine and antagonist of serotonin and histamine2.
  • Inhibits proliferation of hepatocellular carcinoma cells by blocking cell cycle progression through the activation of P38 MAP kinase.
  • Reduces the viability of myeloma and leukemia cell lines.
  • Causes apoptosis and decreases inflammation by disrupting thiol/disulfide balance and enhancing the levels of SIRT1 in C6 glioblastoma cells, and inhibits cell proliferation.
  • Exhibits antitumor activity in urothelial carcinoma cells.
  • Enhances the delayed rectifier outward potassium current in mouse cortical neurons.
  • Increases the excitability of pyramidal neurons in mouse medial prefrontal cortex slices.
  • Inhibits in vitro and in vivo lung metastasis and drives metabolic rewiring.
Catalog No. 50-004-12354 Supplier Medchemexpress LLC Supplier No. HYB116510MM/1ML
Add to Cart
Add to Cart
Cyproheptadine hydrochloride sesquihydrate is an antihistamine and an antagonist of serotonin and histamine2.
  • Antihistamine and antagonist of serotonin and histamine2.
  • Inhibits proliferation of hepatocellular carcinoma cells by blocking cell cycle progression through the activation of P38 MAP kinase.
  • Reduces the viability of myeloma and leukemia cell lines.
  • Causes apoptosis and decreases inflammation by disrupting thiol/disulfide balance and enhancing the levels of SIRT1 in C6 glioblastoma cells, and inhibits cell proliferation.
  • Exhibits antitumor activity in urothelial carcinoma cells.
  • Enhances the delayed rectifier outward potassium current in mouse cortical neurons.
  • Increases the excitability of pyramidal neurons in mouse medial prefrontal cortex slices.
  • Inhibits in vitro and in vivo lung metastasis and drives metabolic rewiring.