Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medisca Cyproheptadine Hydrochloride USP
SDP

Catalog No. 14185530 Shop All Medisca Products
Change view
Click to view available options
Quantity:
5 g
10 g
100 g
1 kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
14-185-530 10 g
14-185-471 5 g
14-185-475 1 kg
14-185-472 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 14-185-530 Supplier Medisca Supplier No. 122601
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Cyproheptadine Hydrochloride USP is a white to slightly yellow, odorless or practically odorless, crystalline powder.

Cyproheptadine Hydrochloride USP is a white to slightly yellow, odorless or practically odorless, crystalline powder.

Alternative name(s): cyproheptadine hydrochloride sesquihydrate

Specifications

Chemical Name or Material Cyproheptadine Hydrochloride USP
Packaging Jar 60ml, White, HDPE, Round, 33-400, TRIVENI-DB, CT
CAS 41354-29-4
Type Drug Standard
Molecular Weight (g/mol) 332.87
Quantity 10 g
Percent Purity 98 to 102 %
Grade USP
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.