Learn More
Selleck Chemical LLC Cyproterone Acetate S2042-10mM/1mL
SDP
Catalog No. 503111126
Encompass_Preferred
Cyproterone acetate is an androgen receptor (AR) antagonist with IC50 of 7 1 nM as well as a weak progesterone receptor agonist with weak pro-gestational and glucocorticoid activity
Catalog No. 50-311-1126 Supplier Selleck Chemical LLC Supplier No. S204210MM/1ML
Cyproterone acetate is an androgen receptor (AR) antagonist with IC50 of 7 1 nM as well as a weak progesterone receptor agonist with weak pro-gestational and glucocorticoid activity