Learn More
Apexbio Technology LLC Cyproterone Acetate 427-51-0 250mg
SDP
Catalog No. 502240702
Encompass_Preferred
Cyproterone Acetate (CAS 427-51-0) is a small-molecule antagonist targeting the androgen receptor (AR) It is designed to inhibit AR activation thereby suppressing androgen-mediated cellular signaling Cyproterone Acetate exerts its biological activity primarily through competitive binding to AR displacing endogenous androgen ligands and blocking receptor activation and downstream signaling In experimental assays Cyproterone Acetate demonstrates AR inhibition with an IC50 value of approximately 7 1 nM At higher concentrations (EC50 around 4 0 M) it may also exhibit partial agonist activity on the AR pathway Based on these pharmacological properties Cyproterone Acetate holds research potential in studies of androgen-dependent biological processes AR signaling mechanisms and hormone-sensitive pathologies such as prostate cancer or androgen-related metabolic conditions
This item is not returnable. View return policy
Catalog No. 50-224-0702 Supplier Apexbio Technology LLC Supplier No. A8404250
Cyproterone Acetate (CAS 427-51-0) is a small-molecule antagonist targeting the androgen receptor (AR) It is designed to inhibit AR activation thereby suppressing androgen-mediated cellular signaling Cyproterone Acetate exerts its biological activity primarily through competitive binding to AR displacing endogenous androgen ligands and blocking receptor activation and downstream signaling In experimental assays Cyproterone Acetate demonstrates AR inhibition with an IC50 value of approximately 7 1 nM At higher concentrations (EC50 around 4 0 M) it may also exhibit partial agonist activity on the AR pathway Based on these pharmacological properties Cyproterone Acetate holds research potential in studies of androgen-dependent biological processes AR signaling mechanisms and hormone-sensitive pathologies such as prostate cancer or androgen-related metabolic conditions