Learn More
Cayman Chemical α-Cyperone, Purity: ≥98%, CAS: 473-08-5, 50 mg
SDP
Catalog No. 502946606
Encompass_Preferred
A sesquiterpenoid with diverse biological activities; inhibits LPS-induced nuclear translocation of p65, as well as LPS-induced production of PGE2 and IL-6, in RAW 264.7 cells; reduces hydrogen peroxide-induced production of ROS and apoptosis in SH-SY5Y cells at 15 and 30 µM; reduces the production of HBsAG in infected HepG2.2.15 cells (IC50 = 274.7 µM)
Catalog No. 50-294-6606 Supplier Cayman Chemical Supplier No. 3429450 MG
A sesquiterpenoid with diverse biological activities; inhibits LPS-induced nuclear translocation of p65, as well as LPS-induced production of PGE2 and IL-6, in RAW 264.7 cells; reduces hydrogen peroxide-induced production of ROS and apoptosis in SH-SY5Y cells at 15 and 30 µM; reduces the production of HBsAG in infected HepG2.2.15 cells (IC50 = 274.7 µM)