Learn More
Contec™ CyQuanol Non-Sterile Disinfectant Solution
BIOPHARMAGREENER_CHOICE
Catalog No. 19161002 Shop All Contec Inc. Products
Change view
Click to view available options
Volume (English):
32 oz.
1 gal.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (English)
19-161-002 1 gal.
19-161-005 32 oz.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 19-161-002 Supplier Contec™ Supplier No. FBC128CQ
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy
Contec™ CyQuanol is a blend of quaternary ammonium and ethyl alcohol that generates low residue.
  • 0.2 micron filtered for the most critical environments
  • Bactericidal, virucidal, fungicidal and tuberculocidal
  • 1-minute contact time makes it easier to achieve dwell time
  • No pre-clean required for this one-step cleaner disinfectant
  • Bleach-free makes it compatible with common cleanroom surfaces
Specifications
Product Type Cleaner/Disinfectant
Sterility Non-sterile
Appearance Clear
Volume (English) 1 gal.
Volume (Metric) 3.8 L