Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Contec™ CyQuanol Sterile Disinfectant Solution
GREENER_CHOICE

Catalog No. 19161004 Shop All Contec Inc. Products
Change view
Click to view available options
Volume (English):
32 oz.
1 gal.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (English)
19-161-004 32 oz.
19-161-003 1 gal.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 19-161-004 Supplier Contec™ Supplier No. SBT32CQ
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Contec™ CyQuanol is a blend of quaternary ammonium and ethyl alcohol that generates low residue.

  • 0.2 micron filtered for the most critical environments
  • Bactericidal, virucidal, fungicidal and tuberculocidal
  • 1-minute contact time makes it easier to achieve dwell time
  • No pre-clean required for this one-step cleaner disinfectant
  • Bleach-free makes it compatible with common cleanroom surfaces

Specifications

Product Type Cleaner/Disinfectant
Sterility Sterile
Appearance Clear
Volume (English) 32 oz.
Volume (Metric) 0.9 L
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.