Learn More
Sigma Aldrich Fine Chemicals Biosciences CyR5 dCTP Cytiva PA55021
Catalog No. 502904452
Encompass
CyDyes are a group of bright fluorescent dyes which can be used in genomic and protein research. Cy(R) dye labelled nucleotides exhibit the following propertiesfor labeling DNA and RNA probes especially for multicolor analysis in microarrays fISH chromosome identification whole chromosome painting karyotyping and gene mapping.CyDye(TM) labeled nucleotides are available in a range of bright intense colors with narrow emission bands well suited for multiplexing within a single sample.
Catalog No. 50-290-4452 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. GEPA55021
CyDyes are a group of bright fluorescent dyes which can be used in genomic and protein research. Cy(R) dye labelled nucleotides exhibit the following propertiesfor labeling DNA and RNA probes especially for multicolor analysis in microarrays fISH chromosome identification whole chromosome painting karyotyping and gene mapping.CyDye(TM) labeled nucleotides are available in a range of bright intense colors with narrow emission bands well suited for multiplexing within a single sample.