Learn More
Sigma Aldrich Fine Chemicals Biosciences CyR5 Mono 5 pack Cytiva PA
Catalog No. 502904344
Encompass
Cy5 Mono-Reactive Dye PackCy5 mono-Reactive Dye Pack. Convenient way to label antibodies proteins drugs and oligonucleotides with CyDye(TM) fluorescent Dyes. . for use in ELISA immunoprecipitation and blotting applications. . Dyes are packaged in premixed amounts and foil-sealed to ensure consistent labelings. Contains derivatized CyDye(TM) that carries only one reactive group on each dye molecule for accurate labeling of amine groups for example on antibodies or oligonucleotides.Each kit contains five vials of dye. Each vial is sufficient for labeling 1 mg of protein or 30 nmol of amino groups.
Catalog No. 50-290-4344 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. GEPA25001
Cy5 Mono-Reactive Dye PackCy5 mono-Reactive Dye Pack. Convenient way to label antibodies proteins drugs and oligonucleotides with CyDye(TM) fluorescent Dyes. . for use in ELISA immunoprecipitation and blotting applications. . Dyes are packaged in premixed amounts and foil-sealed to ensure consistent labelings. Contains derivatized CyDye(TM) that carries only one reactive group on each dye molecule for accurate labeling of amine groups for example on antibodies or oligonucleotides.Each kit contains five vials of dye. Each vial is sufficient for labeling 1 mg of protein or 30 nmol of amino groups.