Learn More
Research Products International Corp Cryogenic Storage Vial, 5ml, Sterile, 100 per Case
SDP
Catalog No. 50476508
Encompass
Convenient 5ml capacity vial is ideal for low temperature freezer storage to -70°C. Vial has a skirted bottom for free standing use and external threading on vial provides a smooth interior surface. Additional features include easy to read white printed graduations and a large marking area. Screw cap features an internal sealing ring for a secure fit. Packaged as 50 vials and caps per bag with 2 bags per case (100 per case).
Catalog No. 50-476-508 Supplier Research Products International Corp Supplier No. 212500
Convenient 5ml capacity vial is ideal for low temperature freezer storage to -70°C. Vial has a skirted bottom for free standing use and external threading on vial provides a smooth interior surface. Additional features include easy to read white printed graduations and a large marking area. Screw cap features an internal sealing ring for a secure fit. Packaged as 50 vials and caps per bag with 2 bags per case (100 per case).