Learn More
Research Products International Corp CryoStore Cryogenic Vials, Conical Bottom, 2.0ml Capacity, 500 per Case
SDP
Catalog No. 50476507
Encompass
CryoStore cryogenic vials are designed for a variety of applications including boiling, freezing and centrifuge applications. Tubes and caps are molded of high density polypropylene and withstand centrifuging speeds up to 17,000 rpm.
Screw caps with 'O' ring feature leak-proof seal that eliminates condensation and prevents sample loss. Tubes are pre-printed with graduations and white marking area. Self standing tubes have interlocking base for use in workstations. Allows for easy one hand capping and uncapping.
Catalog No. 50-476-507 Supplier Research Products International Corp Supplier No. 212434
CryoStore cryogenic vials are designed for a variety of applications including boiling, freezing and centrifuge applications. Tubes and caps are molded of high density polypropylene and withstand centrifuging speeds up to 17,000 rpm.
Screw caps with 'O' ring feature leak-proof seal that eliminates condensation and prevents sample loss. Tubes are pre-printed with graduations and white marking area. Self standing tubes have interlocking base for use in workstations. Allows for easy one hand capping and uncapping.