Learn More
Medchemexpress LLC Cyromazine | 66215-27-8 | 99.8% | 10 G
SDP
Catalog No. 5000412004
Encompass_Preferred
Cyromazine is a triazine insect growth regulator used as an insecticide and an acaricide. It is a cyclopropyl derivative of melamine. This compound works by affecting the nervous system of the immature larval stages of certain insects. Research indicates that Cyromazine can reduce the number of germ cells by interfering with the Ecdysone signaling pathway. It has also been shown to induce renal toxicity, which can be attenuated and restored by green tea extract in rats.
  • Insect growth regulator
  • Insecticide
  • Acaricide
  • Affects the nervous system of immature larval stages of certain insects
  • Reduces germ cells by interfering with the Ecdysone signaling pathway
  • Renal toxicity attenuated by green tea extract in rats
Catalog No. 50-004-12004 Supplier Medchemexpress LLC Supplier No. HYB133110G
Add to Cart
Add to Cart
Cyromazine is a triazine insect growth regulator used as an insecticide and an acaricide. It is a cyclopropyl derivative of melamine. This compound works by affecting the nervous system of the immature larval stages of certain insects. Research indicates that Cyromazine can reduce the number of germ cells by interfering with the Ecdysone signaling pathway. It has also been shown to induce renal toxicity, which can be attenuated and restored by green tea extract in rats.
  • Insect growth regulator
  • Insecticide
  • Acaricide
  • Affects the nervous system of immature larval stages of certain insects
  • Reduces germ cells by interfering with the Ecdysone signaling pathway
  • Renal toxicity attenuated by green tea extract in rats