Learn More
Medchemexpress LLC Cyromazine | 66215-27-8 | 99.8% | 100 G
SDP
Catalog No. 5000413587
Encompass_Preferred
Cyromazine (CGA-72662) is a triazine insect growth regulator used as an insecticide and an acaricide. It is a cyclopropyl derivative of melamine. Cyromazine functions by affecting the nervous system of the immature larval stages of certain insects. It has been shown to reduce the number of germ cells by interfering with the Ecdysone signaling pathway and induces renal toxicity, which can be attenuated and restored by green tea extract in rats.
  • Insect growth regulator
  • Insecticide
  • Acaricide
  • Affects the nervous system of immature larval stages of certain insects
  • Reduces the number of germ cells by interfering with the Ecdysone signaling pathway
  • Induces renal toxicity that can be attenuated by green tea extract in rats
Catalog No. 50-004-13587 Supplier Medchemexpress LLC Supplier No. HYB1331100G
Cyromazine (CGA-72662) is a triazine insect growth regulator used as an insecticide and an acaricide. It is a cyclopropyl derivative of melamine. Cyromazine functions by affecting the nervous system of the immature larval stages of certain insects. It has been shown to reduce the number of germ cells by interfering with the Ecdysone signaling pathway and induces renal toxicity, which can be attenuated and restored by green tea extract in rats.
  • Insect growth regulator
  • Insecticide
  • Acaricide
  • Affects the nervous system of immature larval stages of certain insects
  • Reduces the number of germ cells by interfering with the Ecdysone signaling pathway
  • Induces renal toxicity that can be attenuated by green tea extract in rats