Learn More
Medchemexpress LLC Cyromazine-d4 | 1219804-19-9 | 99.0% | 1 MG
SDP
Catalog No. 5000412495
Encompass_Preferred
Cyromazine-d4 is a deuterium-labeled version of Cyromazine. Cyromazine is a triazine insect growth regulator that functions as an insecticide and acaricide. It is a cyclopropyl derivative of melamine and impacts the nervous system of immature larval stages of certain insects.
  • Used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes of hydrogen, carbon, and other elements can be incorporated into drug molecules for quantitation during drug development.
  • Deuteration has the potential to influence the pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-004-12495 Supplier Medchemexpress LLC Supplier No. HYB1331S1MG
Cyromazine-d4 is a deuterium-labeled version of Cyromazine. Cyromazine is a triazine insect growth regulator that functions as an insecticide and acaricide. It is a cyclopropyl derivative of melamine and impacts the nervous system of immature larval stages of certain insects.
  • Used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes of hydrogen, carbon, and other elements can be incorporated into drug molecules for quantitation during drug development.
  • Deuteration has the potential to influence the pharmacokinetic and metabolic profiles of drugs.