Learn More
Andwin Scientific CYROMOLD 10X10X5
SDP
Catalog No. NC9464348
Encompass_Preferred
The Cryomold molds produce uniformly shaped, flat-surface specimen block when used with O.C.T. (optimum cutting temperature) compound, which is sold separately, for frozen sectioning. This can fit into Cryobar freezing wells of the Tissue Tek II microtome/cryostat with the use of an adapter. This product measures 10 mm x 10 mm x 5 mm.
Catalog No. NC9464348 Supplier Andwin Scientific Supplier No. 25608922
Add to Cart
Add to Cart
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
The Cryomold molds produce uniformly shaped, flat-surface specimen block when used with O.C.T. (optimum cutting temperature) compound, which is sold separately, for frozen sectioning. This can fit into Cryobar freezing wells of the Tissue Tek II microtome/cryostat with the use of an adapter. This product measures 10 mm x 10 mm x 5 mm.