Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Cys-Gly-Tyr-Gly-Pro-Lys-Lys-Lys-Arg-Lys-Val-Gly-Gly | 104914-40-1 | 99.4% | 1377.66 | 5 MG
SDP

Catalog No. 5000397268
Encompass_Preferred

Cys-Gly-Tyr-Gly-Pro-Lys-Lys-Lys-Arg-Lys-Val-Gly-Gly is a 13-mer synthetic peptide containing seven amino acids homologous to SV40 T antigen. It is capable of inducing nuclear transport.

  • Molecular weight: 1377.66
  • Formula: C60H104N20O15S
  • CAS No.: 104914-40-1
  • Appearance: Solid
  • Color: White to off-white
  • Sequence: Cys-Gly-Tyr-Gly-Pro-Lys-Lys-Lys-Arg-Lys-Val-Gly-Gly
  • Sequence shortening: CGYGPKKKRKVGG
  • Purity: 99.43%
  • For research use only
  • Capable of inducing nuclear transport

Catalog No. 50-003-97268 Supplier Medchemexpress LLC Supplier No. HYP38275MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Cys-Gly-Tyr-Gly-Pro-Lys-Lys-Lys-Arg-Lys-Val-Gly-Gly is a 13-mer synthetic peptide containing seven amino acids homologous to SV40 T antigen. It is capable of inducing nuclear transport.

  • Molecular weight: 1377.66
  • Formula: C60H104N20O15S
  • CAS No.: 104914-40-1
  • Appearance: Solid
  • Color: White to off-white
  • Sequence: Cys-Gly-Tyr-Gly-Pro-Lys-Lys-Lys-Arg-Lys-Val-Gly-Gly
  • Sequence shortening: CGYGPKKKRKVGG
  • Purity: 99.43%
  • For research use only
  • Capable of inducing nuclear transport

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.