Learn More
CPC Scientific H-Cys-Leu-Arg-Arg-Ala-Ser-Leu-Gly-OH (trifluoroacetate salt) 1MG
SDP
Supplier:  CPC Scientific PKCP013A
Encompass
ONE-LETTER SEQUENCE: CLRRASLGMOLECULAR FORMULA: C35H66N14O10S1MOLECULAR WEIGHT:875.1STORAGE CONDITIONS: -20 5CRESEARCH AREA: Protein Kinases and PhosphatasesREFERENCES: B.E. Kemp et al., Fed. Proc., 35, 1384, (1976)
Catalog No. 50-211-0166