Learn More
Enzo Life Sciences Cysteinyl Leukotriene ELISA kit (5x96wells)
SDP
Supplier:  Enzo Life Sciences ADI901070
Encompass_Preferred
Highly sensitive ELISA kit enabling detection of LTC4/D4/E4 in 4 hours. The Cysteinyl Leukotriene EIA kit is a colorimetric competitive enzyme immunoassay kit with results in 4 hours. Absorbance is read at 405 nm. Safe non-radioactive measurement of LTC4/D4/E4. Ready-to-use liquid color-coded reagents reduce errors. Sensitivity: 26.6 pg/ml (range 78.1 - 2,500 pg/ml). Use/Stability: Store all components at +4°C, except standard and conjugate at -20°C. Assay Time: 4 hours. Applications: ELISA, Colorimetric detection . Species reactivity: Species independent . Content: GxM Microtiter plate, Conjugate, Antibody, Assay buffer concentrate, Wash buffer concentrate, Standard, pNpp Substrate, Stop solution.
Catalog No. 50-200-6891