Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Cys modifier 1 10mM 1mL | 2374314-08-4 | 381.34 | 1 ML
SDP

Catalog No. 5000439084
Encompass_Preferred

Cys modifier 1 (compound 7) is described as a cysteine-selective protein modifier used for protein bioconjugation. It is a fluorescent carbonylacrylic derivative that bears nitrobenzofurazan, with an excitation wavelength (λex) of 465 nm and an emission wavelength (λem) of 539 nm.

  • Target: Fluorescent dye
  • Pathway: Others
  • Storage: Powder stable for 3 years at -20°C and 2 years at 4°C; In solvent stable for 6 months at -80°C and 1 month at -20°C

Catalog No. 50-004-39084 Supplier Medchemexpress LLC Supplier No. HY11194510MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Cys modifier 1 (compound 7) is described as a cysteine-selective protein modifier used for protein bioconjugation. It is a fluorescent carbonylacrylic derivative that bears nitrobenzofurazan, with an excitation wavelength (λex) of 465 nm and an emission wavelength (λem) of 539 nm.

  • Target: Fluorescent dye
  • Pathway: Others
  • Storage: Powder stable for 3 years at -20°C and 2 years at 4°C; In solvent stable for 6 months at -80°C and 1 month at -20°C

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.