Learn More
Medchemexpress LLC Cys modifier 1 | 2374314-08-4 | 381.34 | 10 MG
SDP
Catalog No. 5000439083
Encompass_Preferred
Cys modifier 1 (compound 7) is a cysteine-selective protein modifier for protein bioconjugation. It is a fluorescent carbonylacrylic derivative bearing nitrobenzofurazan (λex=465nm and λem=539 nm).
  • Cysteine-selective protein modifier
  • Fluorescent (λex=465nm and λem=539 nm)
Catalog No. 50-004-39083 Supplier Medchemexpress LLC Supplier No. HY11194510MG
Cys modifier 1 (compound 7) is a cysteine-selective protein modifier for protein bioconjugation. It is a fluorescent carbonylacrylic derivative bearing nitrobenzofurazan (λex=465nm and λem=539 nm).
  • Cysteine-selective protein modifier
  • Fluorescent (λex=465nm and λem=539 nm)