Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Cys modifier 1 | 2374314-08-4 | 99.0% | 381.34 | 50 MG
SDP

Catalog No. 5000438951
Encompass_Preferred

Cys modifier 1 is a cysteine-selective protein modifier utilized for protein bioconjugation. It is a fluorescent carbonylacrylic derivative that bears nitrobenzofurazan (λex=465nm and λem=539 nm).

  • Cysteine-selective protein modification
  • Fluorescent carbonylacrylic derivative
  • Bears nitrobenzofurazan for fluorescence (λex=465nm and λem=539 nm)
  • Target: Fluorescent dye
  • Pathway: Others

Catalog No. 50-004-38951 Supplier Medchemexpress LLC Supplier No. HY11194550MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Cys modifier 1 is a cysteine-selective protein modifier utilized for protein bioconjugation. It is a fluorescent carbonylacrylic derivative that bears nitrobenzofurazan (λex=465nm and λem=539 nm).

  • Cysteine-selective protein modification
  • Fluorescent carbonylacrylic derivative
  • Bears nitrobenzofurazan for fluorescence (λex=465nm and λem=539 nm)
  • Target: Fluorescent dye
  • Pathway: Others

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.