Learn More
Medchemexpress LLC Cys-PKHB1 TFA | 1487.88 | 10MG
SDP
Catalog No. 5000424780
Encompass_Preferred
Cys-PKHB1 TFA is a peptide conjugated to PKHB1, functioning as a CD47 agonist peptide and a thrombospondin-1 peptide mimic with antitumor effects. It induces mitochondrial alterations, reactive oxygen species generation, and intracellular calcium accumulation, leading to calcium-dependent cell death in breast cancer cells. This compound also activates the immune system in breast cancer cells via immunogenic cell death.
  • Conjugated peptide acting as a CD47 agonist
  • Thrombospondin-1 peptide mimic
  • Exhibits antitumor effects
  • Induces mitochondrial alterations
  • Promotes reactive oxygen species (ROS) generation
  • Causes intracellular calcium (Ca+) accumulation
  • Leads to calcium-dependent cell death in breast cancer cells
  • Activates the immune system through immunogenic cell death
Catalog No. 50-004-24780 Supplier Medchemexpress LLC Supplier No. HYP11205A10MG
Cys-PKHB1 TFA is a peptide conjugated to PKHB1, functioning as a CD47 agonist peptide and a thrombospondin-1 peptide mimic with antitumor effects. It induces mitochondrial alterations, reactive oxygen species generation, and intracellular calcium accumulation, leading to calcium-dependent cell death in breast cancer cells. This compound also activates the immune system in breast cancer cells via immunogenic cell death.
  • Conjugated peptide acting as a CD47 agonist
  • Thrombospondin-1 peptide mimic
  • Exhibits antitumor effects
  • Induces mitochondrial alterations
  • Promotes reactive oxygen species (ROS) generation
  • Causes intracellular calcium (Ca+) accumulation
  • Leads to calcium-dependent cell death in breast cancer cells
  • Activates the immune system through immunogenic cell death